Orders LinkContactUsLinkComponent
QuEChERS standards for AOAC method 2007.01
QuEChERS standards for AOAC method 2007.01
Leverandør:   Restek
QuEChERS standards for AOAC method 2007.01
Leverandør:  Restek
Certifikater
QuEChERS standards for AOAC method 2007.01
QuEChERS standards for AOAC method 2007.01
3 tilgængelige muligheder nedenfor
Product Details & Documents
QuEChERS standards are ready to use for generating test mixes, calibration standards, and spiking experiments.
  • Reliable standards produced according to specifications defined in AOAC official method 2007.01
  • Cost-effective QuEChERS standards can be used without dilutions for greater lab efficiency
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
These standards includes internal standard mix, triphenylphosphate (TPP) solution, and QC spike mix.
QC spike mix contains 27 components: atrazine (1912-24-9); azoxystrobin (131860-33-8); bifenthrin (82657-04-3); carbaryl (sevin) (63-25-2); chlorothalonil (1897-45-6); chlorpyrifos (2921-88-2); chlorpyrifos methyl (5598-13-0); lambda-cyhalothrin (91465-08-6); cyprodinil (121552-61-2); 2,4'-ddd (53-19-0); dichlorvos (ddvp) (62-73-7); endosulfan sulfate (1031-07-8); ethion (563-12-2); imazalil (35554-44-0); imidacloprid (138261-41-3); kresoxim methyl (143390-89-0); linuron (330-55-2); methamidophos (10265-92-6); methomyl (16752-77-5); cis-permethrin (61949-76-6); trans-permethrin (61949-77-7); procymidone (32809-16-8); pymetrozine (123312-89-0); tebuconazole (107534-96-3); thiabendazole (148-79-8); tolylfluanid (731-27-1); trifluralin (1582-09-8).
Product Family Options
Produktinformation
  • Pk Info
    Pakn
    Tilgængelighed
    Price
  • REST31963
    α-BHC-d6 (α-HCH-d6) (86194-41-4), parathion-d10 (350820-04-1), 40 µg/ml each in acetonitrile
    α-BHC-d6 (α-HCH-d6) (86194-41-4), parathion-d10 (350820-04-1), 40 µg/ml each in acetonitrile
    5 ml/ampoule
    1 Ampoul
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 1.560,00 kr
    1.560,00 kr
    1 * 1 Each
       1.560,00 kr
  • REST31964
    Triphenylphosphate solution, 2 µg/ml in acetonitrile:acetic acid (99:1)
    Triphenylphosphate solution, 2 µg/ml in acetonitrile:acetic acid (99:1)
    5 ml/ampoule
    1 Ampoul
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 444,00 kr
    444,00 kr
    1 * 1 Each
       444,00 kr
  • REST31999
    QC spike mix, 40 µg/ml in acetonitrile:acetic acid (99.9:0.1)
    QC spike mix, 40 µg/ml in acetonitrile:acetic acid (99.9:0.1)
    5 ml/ampoule
    1 Ampoul
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5 mL
    1 * 5 mL - 2.030,00 kr
    2.030,00 kr
    1 * 5 mL
       2.030,00 kr