Binocular, magno®
Binocular, 8×, 21 mm
Katalog # ESCH84177821
Leverandør: ESCHENBACH OPTIK
Gå til:
Product Details & Documents
Suitable for a nature getaway, holiday or city break.
- Small-sized, light and handy
- Compact striking design for every occasion
- Multi-layered coating ensuring sharp and true-colour images
Weight: 180 g
Bestillingsoplysninger: Includes a case and carrying cord.
Specifikationer
- Ø:21 mm
- Forstørrelse:8×
- Beskrivelse:Binocular