Orders LinkContactUsLinkComponent
ONPG assay kit
ONPG assay kit
ONPG assay kit, 1 KIT
Catalog # OZBIGO10001
Supplier:   OZ BIOSCIENCES
ONPG assay kit
Catalog # OZBIGO10001
Supplier:  OZ BIOSCIENCES
CAS Number:  
ONPG assay kit
ONPG assay kit
ONPG assay kit, 1 KIT
CAS Number:  
Price
1.482,72 kr
Log in for availability
1 * 1 Each
Product Details & Documents
ONPG assay kit is ideal to quantify high expression level of beta-Gal.
  • Convenient for all transfection assays
  • Suitable for cultured cells and tissues
  • Simple, rapid and easy-to-use
  • Economical
  • Optimised with stable buffers
All the necessary reagents are provided in this assay kit and offer a rapid, simple and sensitive method to quantify spectrophotometrically the enzyme expression level in LacZ transfected cells.
The levels of active β-Gal expression can be quickly measured by its catalytic hydrolysis of ONPG (o-nitrophenyl-beta-D-galactopyranoside) substrate to a bright yellow product (absorbance at 405 to 420 nm). LacZ is one of the most frequently reporter gene used in transfection experiments. The LacZ encoded protein (beta-galactosidase, beta-Gal), is very stable, resistant to proteolytic degradation and easily assayed.
Delivery information: Kit contains 5X lysis buffer (55 ml), standard dillution buffer (55 ml), ONPG substrate solution (55 ml), stop buffer (55 ml) and β-Gal enzyme standard (100 µl).
Caution: Store all reagents at +4 °C except beta Gal enzyme.
Specifications
Specifications
  • Pakn:
    1 KIT
  • Beskrivelse:
    ONPG assay kit