HPLC columns, NUCLEOSIL® OH (Diol)
Supplier: MACHEREY-NAGEL
Certificates
About this item
NUCLEOSIL® OH (Diol) is dihydroxypropyl modified silica phases for HPLC.
- Robust spherical silica gel
- Hydrophilic, polar phase for NP and RP chromatography
- Separation mechanism based on polar interactions (hydrogen bonds)
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
Specifications
- Catalog No:
- MANA720143.46
- MANA721142.30
- pH range:
- 2 - 8
- 2 - 8
- USP code:
- L20
- L20