HPLC columns, NUCLEOSIL® SB
Leverandør: MACHEREY-NAGEL
Om denne vare
NUCLEOSIL® SB is a quaternary ammonium modified silica phase (SAX) for HPLC.
- Robust spherical silica gel
- Anionic phase for ion exchange chromatography
- Separation mechanism based on strong ion exchange interactions
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
Capacity: ~1 meq/g
5 tilgængelige muligheder nedenfor
Product Details & Documents
NUCLEOSIL® SB is a quaternary ammonium modified silica phase (SAX) for HPLC.
- Robust spherical silica gel
- Anionic phase for ion exchange chromatography
- Separation mechanism based on strong ion exchange interactions
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
Capacity: ~1 meq/g
Specifications for Product Family
Specifikationer
- Catalog No:
- MANA720989.46
- MANA720183.46
- MANA720996.46
- MANA720029.46
- MANA721025.30
- Kulstofbelastning:
- 10%
- 10%
- 10%
- 10%
- 10%
- pH-område:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- USP code:
- L14
- L14
- L14
- L14
- L14
Specifikationer
Product Family Options
Analytical columns
Produktinformation
- Kolonne formFasePorestørrelsePartikelstørrelseØ indv.LængdeTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.MANA720989.46Kulstofbelastning10%pH-område2 - 8USP codeL14 - Specifikationer:
More
Specifications
Cat. No.MANA720183.46Kulstofbelastning10%pH-område2 - 8USP codeL14 - Specifikationer:
More
Specifications
Cat. No.MANA720996.46Kulstofbelastning10%pH-område2 - 8USP codeL14 - Specifikationer:
More
Specifications
Cat. No.MANA720029.46Kulstofbelastning10%pH-område2 - 8USP codeL14
Guard column
Produktinformation
- Kolonne formFasePorestørrelsePartikelstørrelseØ indv.LængdeTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.MANA721025.30Kulstofbelastning10%pH-område2 - 8USP codeL14