Orders LinkContactUsLinkComponent
Partial immerstion thermometers
Partial immerstion thermometers
Supplier:   Amarell
Partial immerstion thermometers
Supplier:  Amarell
Certificates
Partial immerstion thermometers
4 Options Available Below
Product Details & Documents
Thermometers are permanently marked.
  • Celsius scale
  • Partial immersion
  • Yellow back
Immersion depth: 76 mm.
Product Family Options
Product Information
  • Område
    Inddeling
    Længde
    Påfyldning
    Neds.- dybde
    Availability
    Price
  • −10...+50 °C
    1,0 °C
    300 mm
    Red special liquid
    76 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 68,20 kr
    68,20 kr
    1 * 1 Items
       68,20 kr
  • −10...+110 °C
    1,0 °C
    300 mm
    Red special liquid
    76 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 67,10 kr
    67,10 kr
    1 * 1 Items
       67,10 kr
  • −10...+150 °C
    1,0 °C
    300 mm
    Red special liquid
    76 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 70,70 kr
    70,70 kr
    1 * 1 Items
       70,70 kr
  • −10...+50 °C
    1,0 °C
    300 mm
    Red special liquid
    76 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 75,00 kr
    75,00 kr
    1 * 1 Items
       75,00 kr