Cleanroom shelf
Cleanroom mini shelf, solid, Rustfrit stål 304, 450×600×260 mm
PALBMB06045-S
: Palbam
Wall mounted mini shelf can be fixed anywhere. Fabrication material is stainless steel 304 with electropolished finish.
- Beskrivelse:Cleanroom mini shelf, solid
- Materiale:Rustfrit stål 304
- B×D×H:450×600×260 mm