SPE columns, acrylamide analysis, CHROMABOND® ABC18
MANA730533
- Kapacitet:6 ml
- Lejevægt:500 mg
- Kolonnemateriale:PP
- Fase:ABC18
- Pakn:30
CHROMABOND® ABC18 is a special phase for SPE analysis of acrylamide in food.
- Special octadecyl phase
- Additional ion exchange functions
Application: Clean-up of acrylamide from ultra-heated starch-containing food, such as potato chips, french fries, crispbread, and cereals.