Empty disposable sample load cartridges, RediSep®
Flashpatron, 5 g
Katalog # HPLCAS-69-3873-235
Leverandør: COLLECTION PREFIX HPLC
Empty disposable sample load cartridges, RediSep®
Katalog # HPLCAS-69-3873-235
Leverandør: COLLECTION PREFIX HPLC
Gå til:
Product Details & Documents
RediSep® empty disposable sample load cartridges improve the resolution of the compound and eliminate reaction byproducts when compared to liquid injection techniques.
Prepare pre-filled solid load cartridges by pipetting the dissolved sample onto the top of the cartridge. Prepare empty solid load cartridges by filling the cartridge with a slurry mixture of the dissolved sample and supporting media. For optimal benefits, remove the solvent by vacuum before placing the cartridge on the purification system.
Specifikationer
- Kolonne form:Flashpatron
- Lejevægt:5 g