Orders LinkContactUsLinkComponent
Magnetic stand
Magnetic stand
Katalog # GENO786-888
Leverandør:   G-Biosciences
Magnetic stand
Katalog # GENO786-888
Leverandør:  G-Biosciences
Magnetic stand
Magnetic stand
Price
2.650,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Magnetic stand is designed for the efficent pull down of magnetic beads in 1,5 and 2 ml micro centrifgue tubes.
  • Suitable for 8×1,5 to 2 ml tubes
  • Pulls beads to tube wall to allow removal of all supernatant
The powerful magnet pulls beads to the side of the tube, allowing for the removal of all the liquid supernatant with no contamination with beads. It can also be used for immunoprecipitations and protein purification analysis.