Wooden applicator
Double swabs, Wood/cotton, 12 mm, 4 mm
Katalog # HERE1032018
Leverandør: HERENZ HEINZ
Gå til:
Product Details & Documents
Wooden applicators with a double cotton bud.
Specifikationer
- Ø:4 mm
- Beskrivelse:Double swabs
- Længde:12 mm
- Skaft/hoved:Wood/cotton