Skip to Header
Skip to Main Content
Skip to Footer
Udvidet søgning
Udvidet søgning
SDS
Certifikater
Producentdata
0
0,00 kr
Order Entry
Log ind
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hjem
Låg
Låg til kromatografirør
PP cap for screw top autosampler vials, AQR™
PP cap for screw top autosampler vials, AQR™
PP cap for screw top autosampler vials, AQR™
MICS9509S-10C-B
:
MICROSOLV
PP cap for screw top autosampler vials, AQR™
MICS9509S-10C-B
:
MICROSOLV
Til:
RSA™ or AQ™ screw top vials with 9-425 threads
Septum:
Non slit, flitted ultra pure silicone/PTFE septa
Lågfarve:
lyseblå
Pakn:
100