Torque wrench
Torque wrench for BPG columns, Rustfrit stål 304, 2 - 20 N
Catalog # 18-0251-37
Supplier: Cytiva
Jump to:
Product Details & Documents
This torque wrench is suitable for assembly and disassembly of BPG columns.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- Beskrivelse:Torque wrench for BPG columns
- Materiale:Rustfrit stål 304
- Seq:30
- Tilspændingsmoment:2 - 20 N