HPLC columns, Cogent UDA™
Supplier: MICROSOLV
Certificates
About this item
These carboxylic acid HPLC columns have a silica hydride surface similar to other TYPE-C™ Silica phases but the uniqueness of Cogent UDA™ is that they can exhibit weak cation exchange properties in addition to RP and ANP retention; a mixed mode is possible with some mobile phases conditions.
- Weak cation
- Mixed mode
A terminal carboxylic acid group contributes some additional selectivity in this column and when the mobile phase is above pH 5,9, the acid group becomes 90% negatively charged and at pH 6,9 is 99% charged and may act as a cation exchanger. Below pH 4,9, the group does not have significant ion exchange properties.
Specifications
- Catalog No:
- 554-6241
- 554-6246
- 554-6245
- 554-6260
- 554-6237
- 554-6269
- 554-6242
- 554-6240
- 554-6268
- 554-6499
- 554-6280
- 554-6486
- 554-6267
- 554-6250
- 554-6279
- 554-6247
- 554-6493
- 554-6236
- 554-6235
- 554-6490
- 554-6489
- 554-6270
- 554-6494
- 554-6278
- 554-6277
- 554-6275
- 554-6274
- 554-6273
- 554-6272
- Kulstofbelastning:
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- 14 - 15%
- Endcapped:
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 340 m²/g
- 390 m²/g
- 340 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 340 m²/g
- 390 m²/g
- 390 m²/g
- 340 m²/g
- 340 m²/g
- 390 m²/g
- 340 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C