HPLC columns, Cogent Bidentate C18™
Supplier: MICROSOLV
Certificates
About this item
Using proprietary bonding technology, the straight chain C18 hydrocarbon is bonded directly to the TYPE-C silica-hydride surface at two separate points of attachment with direct siliconcarbon bonds. This Si-C bond is virtually indestructible and confers great stability and longevity to these columns.
- Suitable for use with mobile phases containing from 100% water to 100% organic solvent
- Very hydrophobic phase
- Columns operate in RP, ONP, and ANP
- Easy gradient analysis
These columns are ideal for generic or USP methods which specify the use of a C18 (or USP L1) column.
Specifications
- Catalog No:
- 554-6202
- 554-6182
- 554-6221
- 554-6181
- 554-6220
- 554-6201
- 554-6178
- 554-6216
- 554-6177
- 554-6219
- 554-6218
- 554-6215
- 554-6176
- 554-6468
- 554-6183
- 554-6469
- 554-6465
- 554-6461
- 554-6214
- 554-6210
- 554-6209
- 554-6211
- 554-6188
- 554-6208
- 554-6191
- 554-6187
- 554-6186
- Kulstofbelastning:
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 20
- 18 - 19
- 20
- 20
- 20
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- 18 - 19
- Endcapped:
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- Ingen
- pH range:
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- 2 - 10
- Surface area:
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 340 m²/g
- 390 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- 390 m²/g
- Temperature limit:
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- 80 °C
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1