Mini mailbox P650 forwarding set white
Mini mailbox P650 forwarding set, white, 240×129×30 mm
Katalog # DAKL520300
Leverandør: DAKLAPACK
Gå til:
Product Details & Documents
The patented MiniMailBox system is approved for use in every country as a recognised safe way of transporting biological substances category B, UN3373 / P650.
- Flap closure
- Made from white cardboard
Commonly used system used by companies, hospitals and testing facilities.
Leveringsinformation: The kit contains the MiniMailBox, Safetybag with bubble liner and absorbing 4 position tube holder.
Specifikationer
- Beskrivelse:Mini mailbox P650 forwarding set, white
- B×D×H:240×129×30 mm