Bottles, narrow neck, square
1000 ml, RD 32, 85×85×221 mm
Katalog # RIXI2-0100-1000-32
Leverandør: BERLIN PACKAGING RIXIUS GMBH
Gå til:
Product Details & Documents
Narrow-neck square bottle made of HDPE.
- Good chemical resistance
- Good temperature resistance
- Robust, natural
Specifikationer
- Gevind:RD 32
- Kapacitet:1000 ml
- B×D×H:85×85×221 mm