Hexamethylenetetramin
Glass bottle for solids, 500 g
Katalog # ROTH4484.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Methenamin
- CAS nummer:100-97-0
- Formel:C₆H₁₂N₄
- MDL nummer:MFCD00006895
- Molecular Weight:140.19 g/mol
- Merck Index:13,05994
- Smeltende Pt:280 °C
- Tæthed:1,33 g/cm³ (20 °C)
- Flash Pt:250 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-905-8
- FN:1328
- ADR:4.1,III
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g