Span® 80, Supelco®
Glass bottle, 50 mL
Katalog # 09569-50ML
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: Arlacel 80, Sorbitan monooleate
- CAS nummer:1338-43-8
- Formel:C₂₄H₄₄O₆
- MDL nummer:MFCD00080948
- Molecular Weight:428.61 g/mol
- Tæthed:1 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ100
- Opbevaringstemperatur:Stue temperatur
- EINECS:215-665-4
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:50 mL