Orders LinkContactUsLinkComponent
5-Bromisoquinolin
5-Bromisoquinolin
Irritant
Supplier:   Cayman Chemical
5-Bromisoquinolin
Supplier:  Cayman Chemical
Certificates
5-Bromisoquinolin
2 Options Available Below
Product Details & Documents
  • CAS Number:34784-04-8
  • Formula:C₉H₆BrN
  • MDL Number:MFCD01646405
  • Molecular Weight:208.06 g/mol
Product Family Options
Product Information
  • Pakning
    Pakn
    Availability
    Price
  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 352,00 kr
    352,00 kr
    1 * 500 mg
       352,00 kr
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 670,00 kr
    670,00 kr
    1 * 1 g
       670,00 kr