Orders LinkContactUsLinkComponent
Microscale tissue grinder kit, KIMBLE®
Microscale tissue grinder kit, KIMBLE®
Microscale tissue grinder kit
Katalog # SCERSP884900-0000
Leverandør:   DWK Life Sciences
Microscale tissue grinder kit, KIMBLE®
Katalog # SCERSP884900-0000
Leverandør:  DWK Life Sciences
Microscale tissue grinder kit, KIMBLE®
Microscale tissue grinder kit, KIMBLE®
Microscale tissue grinder kit
Price
9.960,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The kit contains a variety of the finest microscale tissue grinders suitable for use on tough connective tissue and soft tissue such as brain or liver.
  • Working with micro quantities reduces heat build-up and yields greater enzyme activity
  • Samples can be handled with minimal loss; transfers can often be eliminated
  • HDPE case with die cut foam inserts that protect grinders in transit and in storage
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Leveringsinformation: Kit includes ground glass, smooth glass and PTFE grinders.
Specifikationer
  • Beskrivelse:
    Microscale tissue grinder kit