Sulfiram
Glass bottle for solids, 100 mg
Katalog # PROOBP254
Leverandør: LGC Standards PROMOCHEM
Gå til:
Product Details & Documents
Synonymer: Tetraethylthiuramsulfid
- CAS nummer:95-05-6
- Formel:C₁₀H₂₀N₂S₃
- Molecular Weight:264.48 g/mol
- EINECS:202-387-3
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 mg



