Cetrimoniumbromid
Marketsource
Leverandør: Serva
Gå til:
Om denne vare
Synonymer: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide
- CAS nummer:57-09-0
- Formel:CH₃(CH₂)₁₅N(Br)(CH₃)₃
- MDL nummer:MFCD00011772
- Molecular Weight:364.45 g/mol
- Merck Index:13,02034
- Smeltende Pt:237…243 °C
- Flash Pt:244 °C (closed cup)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-311-3
- FN:3077
- ADR:9,III
2 tilgængelige muligheder nedenfor
Product Details & Documents
Synonymer: N-Cetyl-N,N,N-trimethylammonium bromide, Cetyltrimethylammonium bromide, Hexadecyltrimethylammonium bromide, CTAB, (1-Hexadecyl)trimethylammonium bromide, N,N,N-Trimethylhexadecan-1-aminium bromide
- CAS nummer:57-09-0
- Formel:CH₃(CH₂)₁₅N(Br)(CH₃)₃
- MDL nummer:MFCD00011772
- Molecular Weight:364.45 g/mol
- Merck Index:13,02034
- Smeltende Pt:237…243 °C
- Flash Pt:244 °C (closed cup)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-311-3
- FN:3077
- ADR:9,III