Jump to:
Product Details & Documents
Synonyms: Anhydrone
- CAS Number:10034-81-8
- Formula:Cl₂MgO₈
- MDL Number:MFCD00011102
- Molecular Weight:223.21 g/mol
- Merck Index:13,05702
- Melting Pt:251 °C
- Density:2,21 g/cm³ (20 °C)
- EINECS:233-108-3
- UN:1475
- ADR:5.1,II
Documents
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:454 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



