4-Methylchryene
Glass bottle for solids, 10 mg
Katalog # EHERE20880000
Leverandør: EHRENSTORFER
Gå til:
Product Details & Documents
- CAS nummer:3351-30-2
- Formel:C₁₉H₁₄
- Molecular Weight:242.32 g/mol
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 mg



