2-Methylchryene
Glass bottle for solids, 10 mg
Katalog # PROOBCR-078R
Leverandør: LGC Standards PROMOCHEM
Gå til:
Product Details & Documents
- CAS nummer:3351-32-4
- Formel:C₁₉H₁₄
- Molecular Weight:242.32 g/mol
- Kogende Pt:411 °C (1013 hPa)
- Smeltende Pt:229,5…230 °C
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 mg