Supelco® Organic Reference Standard, PCB No 153
PCB No 153, Analytical standard, Glass bottle for solids, 10 mg
Katalog # 35602-10MG
Leverandør: Merck
Gå til:
Product Details & Documents
- GHS signal: GHS08, GHS09
Advarsel: P260 - P273 - P314 - P391 - P501
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 mg
- Standard:Analytical standard
- Beskrivelse:PCB No 153
- Cat. No.:35602-10MG
- M-Clarity quality level:MQ100
- CAS-nr:35065-27-1
- Molekylvægt:360,88
- MDL No.:MFCD00055535
- Fareklassebeskrivelse:H373 - H410
- Flammepunkt:Not applicable
- Opbevaring betingelser:2...30 °C
- Synonym(s):2,2′,4,4′,5,5′-Hexachlorobiphenyl; 2,2′,4,4′,5,5′-PCB
- Shipping conditions:STD



