Orders LinkContactUsLinkComponent
Phenolphthalein
Glass bottle for solids, 100 g
Katalog # ROTHT126.1
Sundhed
Marketsource
Leverandør:   Roth Carl
Phenolphthalein
Katalog # ROTHT126.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  77-09-8
Phenolphthalein
Phenolphthalein
Glass bottle for solids, 100 g
CAS nummer:  77-09-8
Price
223,28 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
  • CAS nummer:77-09-8
  • Formel:C₂₀H₁₄O₄
  • MDL nummer:MFCD00005913
  • Molecular Weight:318.33 g/mol
  • Merck Index:14,07243
  • Smeltende Pt:258…262 °C
  • Tæthed:1,3 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-004-7
  • NÅ:01-2119498295-24
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g
Specifikationer