Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
4-Hydroxybenzylalkohol

4-Hydroxybenzylalkohol

Glass bottle for solids, 5 g
Katalog # ROTH6257.1
Marketsource
Leverandør:   Roth Carl
4-Hydroxybenzylalkohol
Katalog # ROTH6257.1
Leverandør:  Roth Carl
Marketsource
CAS nummer:  623-05-2
4-Hydroxybenzylalkohol

Glass bottle for solids, 5 g

CAS nummer:  623-05-2
Price
Spørg for pris
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  (4-Hydroxyphenyl)methanol, p-Hydroxybenzyl Alcohol, 4-(Hydroxymethyl)phenol
  • CAS nummer:623-05-2
  • Formel:C₇H₈O₂
  • MDL nummer:MFCD00004658
  • Molecular Weight:124.14 g/mol
  • Kogende Pt:252 °C (1013 hPa)
  • Smeltende Pt:115…118 °C
  • Opbevaringstemperatur:Køleskab
  • EINECS:210-768-0
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.