Orders LinkContactUsLinkComponent
m-Cresol, Supelco®
 :   Merck
m-Cresol, Supelco®
 :  Merck
m-Cresol, Supelco®
m-Cresol, Supelco®
 :  m-Methylphenol
  •  :108-39-4
  •  :CH₃C₆H₄OH
  •  :MFCD00002302
  •  :108.14 g/mol
  •  :13,02604
  •  :202,2 °C (1013 hPa)
  •  :11,5 °C
  •  :1,0336 g/cm³ (20 °C)
  •  :73 °C
  •  :Stue temperatur
  •  :2076
  •  :6.1,II
  •  :M-Clarity™ quality level = MQ100
  •  :203-577-9
Product Family Options
  • Pakning
    Pakn
  • Glass bottle
    1 mL
    1 * 1 mL
    1 * 1 mL - 295,00 kr
    295,00 kr
    1 * 1 mL
       295,00 kr
  • Glass bottle
    5 mL
    1 * 5 mL
    1 * 5 mL - 865,00 kr
    865,00 kr
    1 * 5 mL
       865,00 kr