3'-Chlorpropiophenon 98%
Leverandør: Thermo Fisher Scientific
Om denne vare
Synonymer: m-Chloropropiophenone, 1-(3-Chlorophenyl)propan-1-one
- CAS nummer:34841-35-5
- Formel:C₉H₉ClO
- MDL nummer:MFCD00009925
- Molecular Weight:168.62 g/mol
- Kogende Pt:124 °C (14 mmHg)
- Smeltende Pt:43…47 °C
- Flash Pt:>110 °C
- EINECS:252-242-3
2 tilgængelige muligheder nedenfor
Product Details & Documents
Synonymer: m-Chloropropiophenone, 1-(3-Chlorophenyl)propan-1-one
- CAS nummer:34841-35-5
- Formel:C₉H₉ClO
- MDL nummer:MFCD00009925
- Molecular Weight:168.62 g/mol
- Kogende Pt:124 °C (14 mmHg)
- Smeltende Pt:43…47 °C
- Flash Pt:>110 °C
- EINECS:252-242-3
Documents
ACRO27658
SDS
Specifications for Product Family
Specifikationstestresultater
- Appearance:White to light yellow Crystalline solid
- Infrared spectrum:Conforms
- Melting point:43 to 47 °C
- GC:≥97.5 %