p-Phenylendiamin
Glass bottle for solids, 100 g
Katalog # ROTH4499.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: 1,4-Phenylendiamin
- CAS nummer:106-50-3
- Formel:C₆H₈N₂
- MDL nummer:MFCD00007901
- Molecular Weight:108.14 g/mol
- Merck Index:12,07439
- Kogende Pt:267 °C (1013 hPa)
- Smeltende Pt:139…141 °C
- Tæthed:1,251 g/cm³ (20 °C)
- Flash Pt:156 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-404-7
- FN:1673
- ADR:6.1,III
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g



