Azobenzen 98%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:103-33-3
:
Formula:C₁₂H₁₀N₂
MDL Number:MFCD00003022
Boiling Pt:293
Melting Pt:65…68 °C
Density:1.09 g/ml (25 °C) (lit.)
Molecular Weight:182.22 g/mol
Flash Pt:100 °C (closed cup)
EINECS:203-102-5
UN:3077
ADR:9 III
Merck Index:12,00948
:M-Clarity™ quality level = MQ100
:GHS08
:GHS07
:GHS09
Storage Temperature:Room Temperature
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance (Form):Crystals or Crystals with Chunk(s) or Crystalline Chunks
- Infrared spectrum:Conforms to Structure
- Carbon:77.1 - 81.1%
- Nitrogen:15.0 - 15.8%
- Purity (GC):≥97.5%
Specification Test Results
Frequently Bought Together