Orders LinkContactUsLinkComponent
Diethylphthalat, Supelco®
Diethylphthalat, Supelco®
Glass bottle, 5 mL
Katalog # 53008-5ML-F
Leverandør:   Merck
Diethylphthalat, Supelco®
Katalog # 53008-5ML-F
Leverandør:  Merck
CAS nummer:  84-66-2
Diethylphthalat, Supelco®
Diethylphthalat, Supelco®
Glass bottle, 5 mL
CAS nummer:  84-66-2
Price
995,00 kr
Log ind for tilgængelighed
1 * 5 mL
Product Details & Documents
Synonymer:  Phthalic acid diethyl ester
  • CAS nummer:84-66-2
  • Formel:C₁₂H₁₄O₄
  • MDL nummer:MFCD00009111
  • Molecular Weight:222.24 g/mol
  • Merck Index:12,07527
  • Kogende Pt:298 °C (1013 hPa)
  • Smeltende Pt:–3 °C
  • Tæthed:1,12 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Flash Pt:156 °C (closed cup)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-550-6
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle
  • Pakn:
    5 mL
Specifikationer