Benz[a]anthracen
100 mg, 1
Katalog # 48563.
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: Benz(alpha)anthracen, 1,2-Benzanthracen
- CAS nummer:56-55-3
- Formel:C₁₈H₁₂
- MDL nummer:MFCD00003599
- Molecular Weight:228.29 g/mol
- Merck Index:12,01089
- Kogende Pt:437 °C (1013 hPa)
- Tæthed:1,245 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-280-6
- FN:3077
- ADR:9,III
Specifikationer
Specifikationer
- Pakn:1
- Pk Info:100 mg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



