Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Benz[a]anthracen

Benz[a]anthracen

Glass bottle for solids, 20 mg
Katalog # PROOBCR-271
Sundhed
Miljø
Leverandør:   LGC Standards PROMOCHEM
Benz[a]anthracen
Katalog # PROOBCR-271
CAS nummer:  56-55-3
Benz[a]anthracen

Glass bottle for solids, 20 mg

CAS nummer:  56-55-3
Price
5.270,00 kr
Log ind for tilgængelighed
1 * 20 mg
Product Details & Documents
Synonymer:  Benz(alpha)anthracen, 1,2-Benzanthracen
  • CAS nummer:56-55-3
  • Formel:C₁₈H₁₂
  • MDL nummer:MFCD00003599
  • Molecular Weight:228.29 g/mol
  • Merck Index:12,01089
  • Kogende Pt:437 °C (1013 hPa)
  • Tæthed:1,245 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-280-6
  • FN:3077
  • ADR:9,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    20 mg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.