Supelco® Organic Reference Standard, Anthracene
Anthracene, Analytical standard, Glass bottle for solids, 250 mg
Katalog # 31581-250MG
Leverandør: Merck
Gå til:
Product Details & Documents
- GHS signal: GHS07, GHS09
Advarsel: P264 - P273 - P280 - P305 + P351 + P338 - P337 + P313 - P391
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:250 mg
- Standard:Analytical standard
- Beskrivelse:Anthracene
- MDL No.:MFCD00001240
- Synonym(s):Anthraxcene; Paranaphthalene
- Cat. No.:31581-250MG
- M-Clarity quality level:MQ100
- CAS-nr:120-12-7
- Molekylvægt:178,23
- Kogepunkt:340 °C (lit.)
- Fareklassebeskrivelse:H319 - H410
- Flammepunkt:121,0 °C - closed cup
- Opbevaring betingelser:2...30 °C
- Melting point:210...215 °C (lit.)
- Shipping conditions:STD



