Kanelalkohol
Glass bottle for solids, 100 g
ROTH4905.1
Marketsource
: Cinnamic alcohol, 3-Phenylprop-2-en-1-ol
- :104-54-1
- :C₉H₁₀O
- :MFCD00002921
- :134.18 g/mol
- :13,02325
- :256 °C (1013 hPa)
- :31…34 °C
- :1,04 g/cm³ (25 °C)
- :126 °C
- :Stue temperatur
- :203-212-3
- Pakning:Glass bottle for solids
- Pakn:100 g