Orders LinkContactUsLinkComponent
Isooctan
Isooctan
Flammable
Health
Irritant
Environment
Marketsource
Supplier:   Roth Carl
Isooctan
Supplier:  Roth Carl
Marketsource
Certificates
Isooctan
3 Options Available Below
Product Details & Documents
Synonyms:  Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
  • CAS Number:540-84-1
  • Formula:(CH₃)₂CHCH₂C(CH₃)₃
  • MDL Number:MFCD00008943
  • Molecular Weight:114.23 g/mol
  • Merck Index:13,05212
  • Boiling Pt:99,2 °C (1013 hPa)
  • Melting Pt:–107 °C
  • Density:0,692 g/cm³ (20 °C)
  • Flash Pt:–12 °C
  • Storage Temperature:Stue temperatur
  • EINECS:208-759-1
  • UN:1262
  • ADR:3,II
Product Family Options
Product Information
  • Pakning
    Pakn
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L
    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L
    Specifications:
    Marketsource
    Marketsource
  • 10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L
    Specifications:
    Marketsource
    Marketsource