VWR® Basic, Aprons, PE
Leverandør: VWR Collection
Om denne vare
Disposable aprons made of PE film.
- Free from natural rubber latex
- Available in two different sizes
Thickness: 20 µm
Emballering: 50 pieces per bag, 10 bags per case.
2 tilgængelige muligheder nedenfor
Product Details & Documents
Disposable aprons made of PE film.
- Free from natural rubber latex
- Available in two different sizes
Thickness: 20 µm
Emballering: 50 pieces per bag, 10 bags per case.
Documents
Food contact certificate C Food Contact, BPA, Phthalates Certificate
Yderligere produktdetaljer
TDS VWR® Basic Apron PE 20 µm
Datablad
VWR® Non Regulated Disposables for Production
Produktlitteratur
Specifications for Product Family
Specifikationer
- Catalog No:
- 113-9093
- 113-9094
- Tykkelse:
- 20 µm
- 20 µm