ContactUsLinkComponent
EDTA tetrasodium salt tetrahydrate 98% (by complexometric analysis), Calbiochem®
EDTA tetrasodium salt tetrahydrate 98% (by complexometric analysis), Calbiochem®
Katalog # 34103-500
CAS nummer:  
EDTA tetrasodium salt tetrahydrate 98% (by complexometric analysis), Calbiochem®
Katalog # 34103-500
CAS nummer:  
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g
  • Seq:
    3645
Specifikationer
Om denne vare
 :
 :452.23 g/mol
Merck Index:12,03557
Smeltende Pt:> 300 °C
Tæthed:6,9 g/cm³ (20 °C)
EINECS:200-573-9
 :GHS05
 :GHS07
Opbevaringstemperatur:Stue temperatur
CAS nummer:13235-36-4
Formel:C₁₀H₁₂N₂O₈Na₄·4H₂O
MDL nummer:MFCD00150027