Methyloctanoat
Glass bottle, 10 mL
Katalog # ROTH3331.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
Synonymer: Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate
- CAS nummer:111-11-5
- Formel:C₉H₁₈O₂
- MDL nummer:MFCD00009551
- Molecular Weight:158.24 g/mol
- Kogende Pt:190 °C (1013 hPa)
- Smeltende Pt:–36,7 °C
- Tæthed:0,877 g/cm³ (20 °C)
- Flash Pt:68 °C
- EINECS:203-835-0
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:10 mL
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



