Orders LinkContactUsLinkComponent
Ethylformiat, Supelco®
Glass bottle, 5 mL
  88554-5ML-F
 :   Merck
Ethylformiat, Supelco®
  88554-5ML-F
 :  Merck
 :  109-94-4
Ethylformiat, Supelco®
Ethylformiat, Supelco®
Glass bottle, 5 mL
 :  109-94-4
1.160,00 kr
1 * 5 mL
 :  Formic acid ethyl ester
  •  :109-94-4
  •  :C₃H₆O₂
  •  :MFCD00003294
  •  :74.08 g/mol
  •  :13,03841
  •  :148 °C (1013 hPa)
  •  :–2 °C
  •  :0,9208 g/cm³ (20 °C)
  •  :–20 °C
  •  :Stue temperatur
  •  :1190
  •  :3,II
  •  :M-Clarity™ quality level = MQ100
  •  :203-721-0
  • Pakning:
    Glass bottle
  • Pakn:
    5 mL