Orders LinkContactUsLinkComponent
Acetanilid
Acetanilid
Glass bottle for solids, 100 mg
Katalog # PROOMM0042.10
Irriterende
Leverandør:   LGC Standards PROMOCHEM
Acetanilid
Katalog # PROOMM0042.10
CAS nummer:  103-84-4
Acetanilid
Glass bottle for solids, 100 mg
CAS nummer:  103-84-4
Price
6.410,00 kr
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents
Synonymer:  N-Phenylacetamide
  • CAS nummer:103-84-4
  • Formel:C₈H₉NO
  • MDL nummer:MFCD00008674
  • Molecular Weight:135.17 g/mol
  • Merck Index:13,00051
  • Kogende Pt:304 °C (1013 hPa)
  • Smeltende Pt:115 °C
  • Tæthed:1,22 g/cm³ (20 °C)
  • Flash Pt:174 °C
  • EINECS:203-150-7
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 mg