Orders LinkContactUsLinkComponent
2-Bromethanol analytical standard, Supelco®
1
Catalog # 442334.
Flammable
Irritant
Supplier:   Merck
2-Bromethanol analytical standard, Supelco®
Catalog # 442334.
Supplier:  Merck
CAS Number:  540-51-2
2-Bromethanol analytical standard, Supelco®
2-Bromethanol analytical standard, Supelco®
1
CAS Number:  540-51-2
Price
291,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
  • CAS Number:540-51-2
  • Formula:BrH₂CCH₂OH
  • MDL Number:MFCD00002827
  • Molecular Weight:124.97 g/mol
  • Merck Index:13,03826
  • Boiling Pt:149 °C (1013 hPa)
  • Melting Pt:–80 °C
  • Density:1,76 g/cm³ (20 °C)
  • Flash Pt:40 °C
  • Storage Temperature:Køleskab
  • EINECS:208-748-1
  • UN:2927
  • ADR:6.1,II
Specifications
Specifications
  • Pakn:
    1
Specifications