Orders LinkContactUsLinkComponent
L(-)-Tyrosin
Glass bottle for solids, 1
  CAMBDLM-451-PK
L(-)-Tyrosin
  CAMBDLM-451-PK
 :  60-18-4
L(-)-Tyrosin
Glass bottle for solids, 1
 :  60-18-4
1 * 1 Items
 :  L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH
  •  :60-18-4
  •  :C₉H₁₁NO₃
  •  :MFCD00002606
  •  :181.19 g/mol
  •  :13,09907
  •  :351 °C (1013 hPa)
  •  :285 °C
  •  :1,41 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :200-460-4
  • Pakning:
    Glass bottle for solids
  • Pakn:
    1