L(+)-Leucin
Plastic bottle for solids, 100 g
Catalog # USBIL2020-05-100
Supplier: US Biological
Jump to:
Product Details & Documents
Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
- CAS Number:61-90-5
- Formula:C₆H₁₃NO₂
- MDL Number:MFCD00002617
- Molecular Weight:131.17 g/mol
- Merck Index:14,05451
- Melting Pt:300 °C
- Density:1,165 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:200-522-0
Documents
Specifications
Specifications
- Pakning:Plastic bottle for solids
- Pakn:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



