L(+)-Lithiumlactat
Glass bottle for solids, 10 g
Catalog # RESERES9002L-A102X
Supplier: RESEARCH ORGANICS
Jump to:
Product Details & Documents
Synonyms: Lithium (S)-2-hydroxypropanoate, (S)-(+)-Lactic acid lithium salt, (S)-2-Hydroxypropionic acid Lithium salt, Lithium (S)-2-Hydroxypropionate, L(+)-Lactic acid lithium salt, (S)-(+)-2-Hydroxypropanoic acid lithium salt, (S)-(+)-2-Hydroxypropionic acid lithium salt, Lithium L-Lactate, L-Lactic acid Lithium salt
- CAS Number:27848-80-2
- Formula:C₃H₅LiO₃
- MDL Number:MFCD00065512
- Molecular Weight:96.01 g/mol
- Boiling Pt:200 °C (1013 hPa)
- Melting Pt:75 °C
- Storage Temperature:Stue temperatur
- EINECS:248-692-5
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:10 g



