Hydroxylammoniumchlorid
Plastic bottle for solids, 500 g
Katalog # SERA25513.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Hydroxylamine HCl, Hydroxylamine hydrochloride
- CAS nummer:5470-11-1
- Formel:NH₂OH·HCl
- MDL nummer:MFCD00051089
- Molecular Weight:69.49 g/mol
- Merck Index:13,04853
- Kogende Pt:306 °C (1013 hPa)
- Smeltende Pt:151 °C
- Tæthed:1,67 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:226-798-2
- FN:2923
- ADR:8,III
Specifikationer
Specifikationer
- Pakning:Plastic bottle for solids
- Pakn:500 g