Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Spermidin trihydrochlorid

Spermidin trihydrochlorid

Glass bottle for solids, 5 g
Katalog # SERA35285.02
Marketsource
Leverandør:   Serva
Spermidin trihydrochlorid
Katalog # SERA35285.02
Leverandør:  Serva
Marketsource
CAS nummer:  334-50-9
Spermidin trihydrochlorid

Glass bottle for solids, 5 g

CAS nummer:  334-50-9
Price
1.276,96 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  4-Azoniaoctamethylenediammonium trichloride, 4-Azaoctamethylenediamine trihydrochloride, N1-(3-Aminopropyl)butane-1,4-diamine trihydrochloride, N-(3-Aminopropyl)-1,4-diaminobutane trihydrochloride
  • CAS nummer:334-50-9
  • Formel:C₇H₂₂Cl₃N₃
  • MDL nummer:MFCD00012918
  • Molecular Weight:254.63 g/mol
  • Merck Index:12,08893
  • Smeltende Pt:256…258 °C
  • Opbevaringstemperatur:Fryser
  • EINECS:206-379-0
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.